4-Methylbenzylamine, 98%
4-Methylbenzylamine, 98%
4-Methylbenzylamine, 98%
Thermo Scientific Chemicals

4-Methylbenzylamine, 98%

CAS: 104-84-7 | C8H11N | 121.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
126401000100 gGlass bottle
Catalog number 126401000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS104-84-7
IUPAC Name1-(4-methylphenyl)methanamine
Molecular FormulaC8H11N
InChI KeyHMTSWYPNXFHGEP-UHFFFAOYSA-N
SMILESCC1=CC=C(CN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5330 to 1.5360 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only