Phenylphosphonic acid, 98%
Phenylphosphonic acid, 98%
Phenylphosphonic acid, 98%
Thermo Scientific Chemicals

Phenylphosphonic acid, 98%

CAS: 1571-33-1 | C6H7O3P | 158.09 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1307900252.5 kgPlastic Bottle
Catalog number 130790025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS1571-33-1
IUPAC Namephenylphosphonic acid
Molecular FormulaC6H7O3P
InChI KeyQLZHNIAADXEJJP-UHFFFAOYSA-N
SMILESOP(O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point158°C to 165°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only