2-Thiohydantoin, 98%
2-Thiohydantoin, 98%
2-Thiohydantoin, 98%
Thermo Scientific Chemicals

2-Thiohydantoin, 98%

CAS: 503-87-7 | C3H4N2OS | 116.14 g/mol
Have Questions?
Catalog NumberQuantity
1387600505 g
Catalog number 138760050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS503-87-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to yellow to beige to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only