Rubber Policemen with Angled Edge
Rubber Policemen with Angled Edge
Fisher Scientific™
Rubber Policemen with Angled Edge
Convenient scrapers with angle-shaped edge.
Have Questions?
Catalog NumberFor Use With (Equipment)
14105B
also known as FIS-14105B
0.48 cm dia. Glass Rod
Catalog number 14105B
also known as FIS-14105B
Price (USD)
-
For Use With (Equipment):
0.48 cm dia. Glass Rod
Convenient scrapers with angle-shaped edge
  • Scrape precipitate from container
  • Hard rubber with angled edges
  • Use section of glass rod as handle
Note: Glass rods are not included.
Specifications
For Use With (Application)Designed to scrape precipitate from container by using section of Glass rod as handle
For Use With (Equipment)0.48 cm dia. Glass Rod
Handle TypeGlass Rod (sold separately)
IncludesAngle shaped edge
Product TypePolicemen Scraper
MaterialRubber
Unit SizePack of 72