Digital Titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field.
Fast and Flexible
Interchangeable cartridges allow you to change titrants in seconds. No cleaning or rinsing burets.
Versatile
More than 50 parameter/range combinations are available.
Economical
Concentrated titrants allow 40-100 typical titrations per cartridge.
Specifications
Number of tests: 40 - 100
Platform: Digital Titrator
Specifications
Concentration0.2256 N
For Use With (Application)Digital Titrator Cartridge