3-Bromophenol, 98%
3-Bromophenol, 98%
3-Bromophenol, 98%
Thermo Scientific Chemicals

3-Bromophenol, 98%

CAS: 591-20-8 | C6H5BrO | 173.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 146340100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS591-20-8
IUPAC Name3-bromophenol
Molecular FormulaC6H5BrO
InChI KeyMNOJRWOWILAHAV-UHFFFAOYSA-N
SMILESOC1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Refractive index1.5950 to 1.6010 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid after melting or solid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only