Traceable™ Flip-Stick™ Thermometer
Fisher Scientific™

Traceable™ Flip-Stick™ Thermometer

Multipurpose, fold-out design makes taking probe readings in any position a snap
Have Questions?
Catalog NumberAccuracy
1464845
also known as FISHERTOPSELLING-1464845-EACH
±1°C
Catalog number 1464845
also known as FISHERTOPSELLING-1464845-EACH
Price (USD)
-
Accuracy:
±1°C
Fisher Scientific™ Traceable™ Flip-Stick™ Thermometer with fold-out design makes taking probe readings in any position a snap.
  • Features °F/°C switch
  • Stainless-steel probe
  • Shockproof and drop-proof up to 5 ft.
  • Max/min temperature reading over any period
  • Wide temperature range cover most routine measurements
  • Position probe at any angle
  • Protect probe in fold-up position
  • 0.75 in. Display is easy to read
  • Rapid reading update—every second
  • HOLD switch freezes display to capture a temperature
Specifications
Accuracy±1°C
Diameter (English) Probe0.14 in.
Dimensions (L x W x H)11 x 0.6 in. (28 x 1.5 cm)
For Use With (Application)Food, gases, samples, reagents, water baths and semi solids
IncludesStainless Steel probe
Length (English)11 in.
Length (English) Probe4.5 in.
Length (Metric) Probe11.4 cm
Product TypeDigital Thermometer
Resolution0.1°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
Weight (English)2.4 oz.
Weight (Metric)68 g
Width (English)0.6 in.
CalibratedYes
DisplayDigital
Length (Metric)280 mm
MaterialStainless Steel
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Width (Metric)1.5 cm
Unit SizeEach