Autoclavable Biohazard Waste Bags
Fisher Scientific™

Autoclavable Biohazard Waste Bags

Dual tested compliant.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)Dimensions (L x W)
1482824613 to 16 gal.35 x 25 in.
148282447 to 10 gal.23 x 19 in.
1482824720 to 30 gal.38 x 31 in.
148282458 to 12 gal.30 x 24 in.
1482824844 gal.48 x 37 in.
Catalog number 14828246
Price (USD)
-
Capacity (English):
13 to 16 gal.
Dimensions (L x W):
35 x 25 in.
Dual tested compliant
Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.

  • Integral steam processing indicator that turns brown when autoclaved
  • Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
  • Operating temperature tolerance up to 285degreeFdegree
  • Available in various sizes
  • Bag with black print
  • Gauge: 2mil
Note: Not compliant with Delaware, which requires 3 mil; check state regulations.
Specifications
Dimensions (L x W)35 x 25 in.
Length (Metric)89 cm
Width (Metric)63.5 cm
AutoclavableAutoclavable
Capacity (English)13 to 16 gal.
ColorRed, Black Print
Unit SizeCase of 100