Institutional Trash Can Liners
Fisher Scientific™

Institutional Trash Can Liners

Provide a leak resistant containment solution for non-medical waste disposal
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)ColorMaterialThickness (English)
1482840220 to 30 gal.ClearLLDPE0.65 mil
14828405
also known as FIS-14828405
32 to 33 gal.BrownLLDPE0.65 mil
1482840432 to 33 gal.ClearLLDPE0.65 mil
1482840320 to 30 gal.BrownLLDPE0.65 mil
1482840632 to 33 gal.ClearLLDPE1.15 mil
1482840740 to 45 gal.ClearLLDPE0.80 mil
1482840840 to 45 gal.ClearLLDPE1.15 mil
1482840960 gal.ClearLLDPE0.80 mil
14828410
also known as FIS-14828410
56 gal.ClearLLDPE1.15 mil
148284127 to 10 gal.ClearHDPE0.24 mil
1482841156 gal.BrownLLDPE1.50 mil
1482841420 to 30 gal.ClearHDPE0.40 mil
1482841633 gal.ClearHDPE0.43 mil
14828413
also known as FIS-14828413
12 to 16 gal.ClearHDPE0.31 mil
1482841733 gal.ClearHDPE0.63 mil
1482841840 to 45 gal.ClearHDPE0.47 mil
1482841940 to 45 gal.ClearHDPE0.63 mil
1482842060 gal.ClearHDPE0.63 mil
14828421
also known as FIS-14828421
56 gal.ClearHDPE0.47 mil
Catalog number 14828402
Price (USD)
-
Capacity (English):
20 to 30 gal.
Color:
Clear
Material:
LLDPE
Thickness (English):
0.65 mil
Provide a leak resistant containment solution for non-medical waste disposal

  • For collecting and containing refuse
  • Available in both linear low density (LLDPE) and high density (HDPE) polyethylene film
  • Sold in flat packs for easy dispensing
  • Provide a cost-effective solution for every application

Specifications
Capacity (English)20 to 30 gal.
Capacity (Metric)75 to 113 L
For Use With (Application)Collecting and containing refuse, non medical waste disposal
ColorClear
MaterialLLDPE
Product TypeInstitutional Trash Can Liner
Thickness (English)0.65 mil
Unit SizeCase of 500