Product Image
Thermo Scientific Chemicals

Methyl phenylacetate, 99+%

CAS: 101-41-7 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
Catalog number 148440050
Price (USD)
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS101-41-7
IUPAC Namemethyl 2-phenylacetate
Molecular FormulaC9H10O2
InChI KeyCRZQGDNQQAALAY-UHFFFAOYSA-N
SMILESCOC(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.5060 to 1.5080 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only