Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Ninhydrin, spectrophotometric grade
Thermo Scientific Chemicals

Ninhydrin, spectrophotometric grade

CAS: 485-47-2 | C9H6O4 | 178.143 g/mol
Have Questions?
Catalog NumberQuantityPackaging
165875000500 gGlass bottle
Catalog number 165875000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS485-47-2
IUPAC Name2,2-dihydroxy-2,3-dihydro-1H-indene-1,3-dione
Molecular FormulaC9H6O4
InChI KeyFEMOMIGRRWSMCU-UHFFFAOYSA-N
SMILESOC1(O)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Suitability(for amino acid analysis) Passes test
Appearance (Color)White to light yellow
Infrared spectrumConforms
pH4.0 to 5.6 (1 % in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only