2,2,6,6-Tetramethyl-3,5-heptanedione, 98%
2,2,6,6-Tetramethyl-3,5-heptanedione, 98%
2,2,6,6-Tetramethyl-3,5-heptanedione, 98%
Thermo Scientific Chemicals

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%

CAS: 1118-71-4 | C11H20O2 | 184.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1683200505 gGlass bottle
Catalog number 168320050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1118-71-4
IUPAC Name2,2,6,6-tetramethylheptane-3,5-dione
Molecular FormulaC11H20O2
InChI KeyYRAJNWYBUCUFBD-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)CC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4579 to 1.4599 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only