alpha-Toluenesulfonyl chloride, 98%
alpha-Toluenesulfonyl chloride, 98%
alpha-Toluenesulfonyl chloride, 98%
Thermo Scientific Chemicals

alpha-Toluenesulfonyl chloride, 98%

CAS: 1939-99-7 | C7H7ClO2S | 190.64 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1704900505 gGlass bottle
Catalog number 170490050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1939-99-7
IUPAC Namephenylmethanesulfonyl chloride
Molecular FormulaC7H7ClO2S
InChI KeyOAHKWDDSKCRNFE-UHFFFAOYSA-N
SMILESClS(=O)(=O)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point90°C to 94°C
Titration Argentometric>=97.5 % (ex Cl)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only