Specifications
AutoclavableNot Autoclavable
Certifications/ComplianceISO 10993-5, HPLC certified, USP Biological Reactivity tests for plastics Class VI, Title 21 of the Code of Federal Regulations (CFR), Section 210.3(b)(6) and 211.72.
ColorBrown
DescriptionFlow Rate: 600 mL/min with water (at 1 bar (14.5 psi))
Diameter (Metric)28 mm
Dimensions (L x W x H)Housing: 33 x 24 mm
Filtration Area6.2 cm2
For Use With (Application)Buffer & Media
Hold up Volume (After Air Purge)100 to 150 μL before bubble point (0.4 bar)
Housing MaterialAcrylic Resin MBS (House)
HydrophobicityHydrophilic
Inlet StyleFemale Luer Lock
No. per Pack50/Pk.
Outlet StyleMale Luer Lock
Pressure Rating4.5 bar (65 psi)
Product LineMinisart
Temperature (English) Max.140°F
Temperature (Metric) Max60°C
TypeSyringe Filter - Type 17594
Volume (Metric)Up to 200 mL
Material (Membrane)SFCA
Pore Size5 μm
SterilitySterile
Unit SizeEach