2,6-Dichloro-3-nitropyridine, 90%, Tech.
2,6-Dichloro-3-nitropyridine, 90%, Tech.
2,6-Dichloro-3-nitropyridine, 90%, Tech.
Thermo Scientific Chemicals

2,6-Dichloro-3-nitropyridine, 90%, Tech.

CAS: 16013-85-7 | C5H2Cl2N2O2 | 192.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 186000100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16013-85-7
IUPAC Name2,6-dichloro-3-nitropyridine
Molecular FormulaC5H2Cl2N2O2
InChI KeySHCWQWRTKPNTEM-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
GC>=88 %
Appearance (Color)Yellow to orange-brown
Water=<10 % (K.F.)
Melting point58°C to 68°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only