Thermo Scientific Chemicals

Methoxyacetic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 186770250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methoxyacetic acid, 97+%, Thermo Scientific Chemicals

Catalog number: 186770250
25 mL, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
625-45-6
IUPAC Name
2-methoxyacetic acid
Molecular Formula
C3H6O3
InChI Key
RMIODHQZRUFFFF-UHFFFAOYSA-N
SMILES
COCC(O)=O
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4150 to 1.4180 (20°C, 589 nm)
Appearance (Form)
Liquid
Titration with NaOH
97.0 to 103.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text