L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

L(+)-Arginine, 98.5-101.5%, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2066-061 kg
2066-0712 kg
2066-0950 kg
Catalog number 2066-06
Price (USD)
-
Quantity:
1 kg
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
This product is GMP Manufactured and meets USP, JP, BP, EP and specifications.
Specifications
Boiling Point368°C (1013 hPa)
CAS74-79-3
Chemical Name or MaterialL(+)-Arginine
ColorColorless
Melting Point238°C
Molecular FormulaC6H14N4O2
PackagingPoly bottle, tamper-evident cap
SynonymL-Arginine, H-L-Arg-OH, Arginine, (S)-(+)-Arginine, (S)-2-Amino-5-guanidinopentanoic acid
GradeMulti-Compendial/USP
Molecular Weight (g/mol)174.2
Percent Purity98.5 to 101.5%
Quantity1 kg
Unit SizeEach