Universal Test Tube Plug Caps
Universal Test Tube Plug Caps
Fisherbrand™

Universal Test Tube Plug Caps

For plastic or glass test tubes.
Have Questions?
Catalog NumberColor
21-235-720Yellow
Catalog number 21-235-720
Price (USD)
-
Color:
Yellow
For plastic or glass test tubes.

Innovative flanged-thread design allows cap to descend into the tube until a friction fit is secure, and plug is stationary.

  • Fit tubes with 12 to 16mm diameter
  • Hollow top finish with ribs for easy retrieval

Specifications
For Use With (Equipment)Fits Tubes with Inner Diameters of 12 to 16 mm
No. per Case1000
Quantity1000/Cs.
ColorYellow
MaterialLDPE
Product TypeTest Tube Plug Cap
SterilityNon-sterile
Unit SizeEach