Wheaton™ Serum Vials
Wheaton™ Serum Vials
DWK Life Sciences

Wheaton™ Serum Vials

Available as clear and amber vials
Have Questions?
Catalog NumberVolume (Metric)ColorDiameter (Metric) Outer
22368610.0 mLClear24 mm
Catalog number 223686
Price (USD)
-
Volume (Metric):
10.0 mL
Color:
Clear
Diameter (Metric) Outer:
24 mm
Available as clear and amber vials

Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.

  • Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
  • Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
  • Withstand extreme temperature variations
  • Necks finished for aluminum crimp seals
  • Blowback feature of neck and specially designed bottom radius for lyophilization applications
  • Packaged in shrink-wrapped modules to reduce particulate contamination

Lyophilization, packaging, and sampling

Specifications
AutoclavableAutoclavable
Certifications/ComplianceASTM E438, USP Type I, Type I, Class A
Diameter (English) Inner0.51 in.
Diameter (Metric) Inner13 mm
For Use With (Application)For lyophilization, packaging, sampling and freeze drying applications
Height (Metric)50 mm
Product TypeSerum Vial
Volume (English)2.7 dr.
Volume (Metric) Total10 mL
Closure TypeCrimp Cap
ColorClear
Diameter (Metric) Outer24 mm
MaterialBorosilicate Glass
Product LineWheaton
Volume (Metric)10.0 mL
Unit SizeEach