Product Image
Hach Pacific Pty Ltd

Nitrogen-Nitrate Reagent Set, LR, Cadmium Reduction

For determination of Nitrate Nitrogen by the Cadmium Reduction method.
Have Questions?
Catalog NumberQuantity
242980010 mL
Catalog number 2429800
Price (USD)
-
Quantity:
10 mL
For determination of Nitrate Nitrogen by the Cadmium Reduction method. Hach Method 8192. Range: 0.01 - 0.50 mg/L NO3-N. Sample Size: 10 mL. Set contains NitraVer™ 6 and NitriVer™ 3 Nitrite Reagent Powder Pillows for 100 tests.
    Specifications

  • Instrument: All except DR/3000, DR/2010, DR/2000, PC II
  • Method: 8192
  • Method Name: Cadmium Reduction
  • Number of tests: 100
  • Parameter: Nitrate, Nitrogen
  • Platform: Powder Pillows
  • Range: 0.01 - 0.50 mg/L NO₃-N
Specifications
Quantity10 mL
Range0.01 to 0.50 mg/L NO3-N
TypePowder Pillows
Volume (Metric)10 mL
Unit SizeEach