Methanol, UNIVAR™, AJAX FineChem™™
AJAX FineChem™

Methanol, UNIVAR™, AJAX FineChem™™

Methanol UNIVAR 10L Conforms to ACS Plastic Bottle CAS No. 67-56-1
Have Questions?
Catalog number 318-10L
Price (USD)
-
Methanol UNIVAR 10L Conforms to ACS Plastic Bottle CAS No. 67-56-1
Specifications
Boiling Point65.4°C
CAS67-56-1
Chemical Name or MaterialMethanol
IUPAC Namemethanol
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
MDL NumberMFCD00004595
Molecular FormulaCH4O
PackagingPlastic
SMILESCO
GradeAnalytical
Molecular Weight (g/mol)32.04
Quantity10 L
Unit SizeEach