Chloromethyl chlorosulfate, 97%
Chloromethyl chlorosulfate, 97%
Chloromethyl chlorosulfate, 97%
Thermo Scientific Chemicals

Chloromethyl chlorosulfate, 97%

CAS: 49715-04-0 | CH2Cl2O3S | 164.984 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 330800050
Price (USD)
-
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS49715-04-0
IUPAC Namechloro({[(chlorooxy)sulfonyl]oxy})methane
Molecular FormulaCH2Cl2O4S
InChI KeyZFEQQZXFULOKEK-UHFFFAOYSA-N
SMILESClCOS(=O)(=O)OCl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Refractive index1.4460 to 1.4480 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only