HPLC Qualification Kits for UltiMate™ 3000 Systems
Actual product may vary
Thermo Scientific™

HPLC Qualification Kits for UltiMate™ 3000 Systems

HPLC Qualification Kits are for system validation of UltiMate 3000 LC and UHPLC systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
3321.0010AIQ Standards for Systems with UV Detector (Caffeine/Uracil)
3323.0010OP/PQ Standards Refill Kit
4832.5000AQualification Kit (OQ/PQ) - Manual Injector
5705.0050AColumn Thermostat PQ Kit
4832.5010AQualification Kit (OQ/PQ) - Autosampler
3325.0010OQ/PQ Standards for RI-Detector
Catalog number 3321.0010A
Price (USD)
-
Description:
IQ Standards for Systems with UV Detector (Caffeine/Uracil)

Choose Thermo Scientific™ Dionex™ UltiMate™ HPLC Qualification Kits for system validation of UltiMate 3000 LC and UHPLC systems.

The modules of UltiMate 3000 RSLCnano system can be arranged in different configurations for various applications.
  • Components include fittings, capillaries and columns, but vary by application
Specifications
DescriptionIQ Standards for Systems with UV Detector (Caffeine/Uracil)
For Use With (Equipment)Dionex UltiMate 3000 Series
Product LineUltiMate
Unit SizeEach