1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

CAS: 22572-40-3 | C9H20IN3 | 297.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
33412010010 gGlass bottle
Catalog number 334120100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS22572-40-3
IUPAC Name(3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular FormulaC9H20IN3
InChI KeyAGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES[I-].CCN=C=NCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point92°C to 97°C
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only