6-Aminonicotinic acid methyl ester, 97%
6-Aminonicotinic acid methyl ester, 97%
6-Aminonicotinic acid methyl ester, 97%
Thermo Scientific Chemicals

6-Aminonicotinic acid methyl ester, 97%

CAS: 36052-24-1 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 366810050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36052-24-1
IUPAC Name2-amino-5-(methoxycarbonyl)pyridin-1-ium
Molecular FormulaC7H9N2O2
InChI KeyJACPDLJUQLKABC-UHFFFAOYSA-O
SMILESCOC(=O)C1=CC=C(N)[NH+]=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to yellow to light beige to pink
Infrared spectrumConforms
Melting point155°C to 160°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only