9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%
Thermo Scientific Chemicals

9,9-Dimethyl-4,5-bis(diphenylphosphino)xanthene, 98%

CAS: 161265-03-8 | C39H32OP2 | 578.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 378060050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS161265-03-8
IUPAC Name[5-(diphenylphosphanyl)-9,9-dimethyl-9H-xanthen-4-yl]diphenylphosphane
Molecular FormulaC39H32OP2
InChI KeyCXNIUSPIQKWYAI-UHFFFAOYSA-N
SMILESCC1(C)C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point226°C to 230°C
Titration with HClO497.5 to 102.5 %
Appearance (Color)White to light yellow
Appearance (Form)powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only