5-Amino-1-pentanol, 50 wt.% aqueous solution
5-Amino-1-pentanol, 50 wt.% aqueous solution
5-Amino-1-pentanol, 50 wt.% aqueous solution
Thermo Scientific Chemicals

5-Amino-1-pentanol, 50 wt.% aqueous solution

CAS: 2508-29-4 | C5H13NO | 103.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
Glass bottle
Catalog number 387918000
Price (USD)
-
Quantity:
800 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2508-29-4
IUPAC Name5-hydroxypentan-1-aminium
Molecular FormulaC5H14NO
InChI KeyLQGKDMHENBFVRC-UHFFFAOYSA-O
SMILES[NH3+]CCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Solution
Appearance (Color)Clear colorless to light yellow
Titration with HCl48.0 to 52.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only