N,N-Diethyl-p-phenylenediamine sulfate, 99%
N,N-Diethyl-p-phenylenediamine sulfate, 99%
N,N-Diethyl-p-phenylenediamine sulfate, 99%
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 99%

N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2 | CAS: 6283-63-2 | C10H16N2·H2SO4 | 262.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 395002500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6283-63-2
IUPAC NameN1,N1-diethylbenzene-1,4-diamine; sulfuric acid
Molecular FormulaC10H18N2O4S
InChI KeyAYLDJQABCMPYEN-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CCN(CC)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals and/or lumps
Infrared spectrumConforms
Melting point184°C to 187°C
Titration with NaOH>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only