5-Aminoimidazole-4-carboxamide, 95%
5-Aminoimidazole-4-carboxamide, 95%
5-Aminoimidazole-4-carboxamide, 95%
Thermo Scientific Chemicals

5-Aminoimidazole-4-carboxamide, 95%

CAS: 360-97-4 | C4H6N4O | 126.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3975500505 gGlass bottle
Catalog number 397550050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS360-97-4
IUPAC Name4-amino-1H-imidazole-5-carboxamide
Molecular FormulaC4H6N4O
InChI KeyDVNYTAVYBRSTGK-UHFFFAOYSA-N
SMILESNC(=O)C1=C(N)N=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to purple
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only