2-Bromo-5-fluoropyridine, 98%, Thermo Scientific Chemicals
2-Bromo-5-fluoropyridine, 98%, Thermo Scientific Chemicals
2-Bromo-5-fluoropyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-5-fluoropyridine, 98%, Thermo Scientific Chemicals

CAS: 41404-58-4 | Molecular Formula: C5H3BrFN | Molecular Weight: 175.98 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3979700505 gGlass bottle
Catalog number 397970050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS41404-58-4
IUPAC Name2-bromo-5-fluoropyridine
Molecular FormulaC5H3BrFN
InChI KeyUODINHBLNPPDPD-UHFFFAOYSA-N
SMILESFC1=CC=C(Br)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless white to light yellow
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=97.5 %

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only