Ethyl isocyanatoacetate, 98%
Ethyl isocyanatoacetate, 98%
Ethyl isocyanatoacetate, 98%
Thermo Scientific Chemicals

Ethyl isocyanatoacetate, 98%

CAS: 2949-22-6 | C5H7NO3 | 129.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 410150050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2949-22-6
IUPAC Nameethyl 2-isocyanatoacetate
Molecular FormulaC5H7NO3
InChI KeyDUVOZUPPHBRJJO-UHFFFAOYSA-N
SMILESCCOC(=O)CN=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Refractive index1.4210 to 1.4230 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only