1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%
Thermo Scientific Chemicals

1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%

CAS: 521-24-4 | C10H5NaO5S | 260.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
41540010010 gGlass bottle
Catalog number 415400100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS521-24-4
IUPAC Namesodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate
Molecular FormulaC10H5NaO5S
InChI KeyUBLXEEBHYISRFM-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Powder
Infrared spectrumConforms
Titration Iodometric>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only