Poly(ethylene Glycol), Average M.W. 8000
Poly(ethylene Glycol), Average M.W. 8000
Poly(ethylene Glycol), Average M.W. 8000
Thermo Scientific Chemicals

Poly(ethylene Glycol), Average M.W. 8000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
41805010010 kgPlastic drum
Catalog number 418050100
Price (USD)
-
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (K.F.)
Hydroxyl value12 to 16 mg KOH/g
Infrared spectrumConforms
Appearance (Color)White
Appearance (Form)Flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only