Methyl picolinate, 98%
Methyl picolinate, 98%
Methyl picolinate, 98%
Thermo Scientific Chemicals

Methyl picolinate, 98%

CAS: 2459-07-6 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 438140050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS2459-07-6
IUPAC Namemethyl pyridine-2-carboxylate
Molecular FormulaC7H7NO2
InChI KeyNMMIHXMBOZYNET-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange
GC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only