4-Methoxy-N-methylbenzylamine, 95%
4-Methoxy-N-methylbenzylamine, 95%
4-Methoxy-N-methylbenzylamine, 95%
Thermo Scientific Chemicals

4-Methoxy-N-methylbenzylamine, 95%

CAS: 702-24-9 | C9H13NO | 151.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 451730100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS702-24-9
IUPAC Name[(4-methoxyphenyl)methyl](methyl)amine
Molecular FormulaC9H13NO
InChI KeyAIJFPNKGGAPZFJ-UHFFFAOYSA-N
SMILESCNCC1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5290 to 1.5310 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only