VACUETTE™ Z No Additive Blood Collection Tubes
Greiner Bio-One™

VACUETTE™ Z No Additive Blood Collection Tubes

VACUETTE™ Z No Additive Collection Tubes are used in determinations for clinical chemistry and hormones, microbiological serology, immunology, TDM and crossmatching. These white top no additive blood tubes are typically used as discard tubes or as an alternative to serum tubes with a clot activator. They contain no chemical additives, are evacuated and sterile.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
454001TSPK
also known as 454001
4 mL
454001TS
also known as 454001
4 mL
Catalog number 454001TSPK
also known as 454001
Price (USD)
-
Capacity (Metric):
4 mL
VACUETTE™ Z No Additive Collection Tubes are used in determinations for clinical chemistry and hormones, microbiological serology, immunology, TDM and crossmatching. These white top no additive blood tubes are typically used as discard tubes or as an alternative to serum tubes with a clot activator. They contain no chemical additives, are evacuated and sterile.
  • Tubes are supplied in packs of 50 pieces
  • For venous blood collection
  • Evacuated and are sterile
  • Made out of virtually unbreakable PET plastic
  • They provide improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Contain no additive in the tube. After one hour the blood in the tube is completely coagulated
  • CE marked
Specifications
Closure ColorWhite
Diameter (Metric) Inner13 mm
Diameter (Metric) Outer13 mm
For Use With (Application)Blood collection
For Use With (Equipment)VACUETTE™ Blood Collection Needles
Height (Metric)75 mm
Length (Metric)75 mm
Tube Size13 x 75 mm
AdditiveNo Additive
Capacity (Metric)4 mL
Closure TypePremium Safety Twist Cap
Product TypeBlood Collection Tube
SterilitySterile
Unit SizeEach