FlaskScrubber™ / FlaskScrubber™ Vantage Glassware Washer Accessory, Upper Spindle Rack
Fisher Scientific™
FlaskScrubber™ / FlaskScrubber™ Vantage Glassware Washer Accessory, Upper Spindle Rack
Outfit your glassware washer with a Labconco Upper Spindle Rack to provide a secure fit during direct injection cleaning.
Have Questions?
Catalog number 4595600
also known as LABCONCO-4595600-EACH
Price (USD)
-
Outfit your glassware washer with a Labconco Upper Spindle Rack to provide a secure fit during direct injection cleaning.
  • Upper Spindle Rack provides direct injection cleaning through the spindles
  • Constructed of Type 304 stainless steel
  • Spindles may be removed and plugged to accommodate accessory inserts (sold separately)
  • Spindle dimensions: 5.25 high x 1/4 in. OD
  • Accommodates glassware up to 8.5 in. high
  • Rack designed for FlaskScrubber and FlaskScrubber Vantage Series Glassware Washers provides forced air drying through the spindles and includes side rails for tank side wall installation
Specifications
Dimensions (L x W x H)53.08 x 50.8 x 18.28cm (20.9 x 20 x 7.2 in.)
For Use With (Equipment)FlaskScrubber™, FlaskScrubber™ Vantage series glassware washer
No. of Pins30
Rack TypeUpper Spindle
Weight (English) Shipping20 lb.
Weight (Metric) Shipping9 kg
MaterialStainless Steel (304)
Product TypeRack
Unit SizeEach