UltiMate™ 3000 Electrochemical Detector Sensors
Product Image
Thermo Scientific™
UltiMate™ 3000 Electrochemical Detector Sensors
Choose either a coulometric or an amperometric analytical sensor for your electrochemical detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
6070.24006011RS Ultra Coulometric Analytical Sensor (Dual Electrode)UltiMate 3000 Colometric Detector
6070.30006041RS Ultra Amperometric Analytical Sensor with Accessories and GasketsUltiMate 3000 Amperometric Detector
6070.21006020RS Omni Coulometric Sensor (Single Electrode)UltiMate 3000 Coulometric Detector
Catalog number 6070.2400
Price (USD)
-
Type:
6011RS Ultra Coulometric Analytical Sensor (Dual Electrode)
For Use With (Equipment):
UltiMate 3000 Colometric Detector
Choose either a coulometric or an amperometric analytical Thermo Scientific™ Dionex™ UltiMate™ 3000 Electrochemical Detector Sensor for your electrochemical detector. They offer both high sensitivity and selectivity for your target analytes.
Specifications
Detection ModeDC
For Use With (Application)Coulometric Detection
For Use With (Equipment)UltiMate 3000 Colometric Detector
No. of Channels2
Reference ElectrodePd
Working ElectrodesPG
Detector TypeElectrochemical (ECD)
Type6011RS Ultra Coulometric Analytical Sensor (Dual Electrode)
Unit SizeEach