Vanquish™ Flow Cells for Vanquish™ Diode Array Detector FG
Thermo Scientific™
Vanquish™ Flow Cells for Vanquish™ Diode Array Detector FG
Choose from a wide portfolio of flow cells for the Thermo Scientific™ Vanquish™ Diode Array Detector (DAD) FG to match your application-related requirements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
6083.0580Flow Cell
6083.0510Standard Flow Cell
6083.0520Semi-Analytical Flow Cell
6083.0530Semi-Micro Flow Cell
6083.0540Standard Biocompatible Flow Cell
6083.0550Semi-Micro Biocompatible Flow Cell
6083.0570Diagnostic Cell
Catalog number 6083.0580
Price (USD)
-
Type:
Flow Cell
Choose from a wide portfolio of Thermo Scientific™ Vanquish™ Flow Cells for the Thermo Scientific™ Vanquish™ Diode Array Detector FG. There's a flow cell to match your application-related requirements including low-dispersion for UHPLC separations, high-pressure resistance for LC-MS or fractionation, and compatibility with the analysis of biomolecules.
Great flow cells for a wide range of applications
12 MPa (120 bar, 1740 psi) flow cell pressure limit:
  • Standard flow cell
  • Semi-analytical flow cell
  • Semi-micro flow cells (biocompatible and SST version available)
5 MPa (50 bar, 720 psi) flow cell pressure limit:
  • Standard biocompatible flow cell
Specifications
MaterialInert
Path Length7 mm
For Use With (Equipment)Vanquish Diode Array Detector FG
Product LineVanquish
TypeFlow Cell
Unit SizeEach