UltiMate™ AFC-3000 Collection Racks
Actual product may vary
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
Thermo Scientific™
UltiMate™ AFC-3000 Collection Racks
Find the right rack for collecting your HPLC sample fractions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6702.004040 Positions, 20 mL (21 mm O.D.)
6702.002121 Positions, 50 mL (33 mm O.D.)
6702.002424 Positions; 30 mL (26 mm O.D.)
6702.006060 Positions, 14 mL (15 mm O.D.)
6702.009090 Positions, 8 mL (13 mm O.D.)
Catalog number 6702.0040
Price (USD)
-
Sample Capacity:
40 Positions, 20 mL (21 mm O.D.)
Collect your samples for further analysis or storage in Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Collection Racks. The racks are available for a range of tube sizes from 8-50mL. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Specifications
For Use With (Equipment)UltiMate AFC-3000 Fraction Collector
Sample Capacity40 Positions, 20 mL (21 mm O.D.)
TypeAFC-3000 Collection Rack
Unit SizeEach