UltiMate™ ACC-3000T Vial Adapters
Actual product may vary
Thermo Scientific™
UltiMate™ ACC-3000T Vial Adapters
Allow the use of 2 mL vials or uncapped 10 mL vials in your Thermo Scientific™ Dionex™ UltiMate™ ACC-3000T column compartment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
6820.2402Adapter for Vial Pusher
6820.4092Adapter for 5-Position Holder
Catalog number 6820.2402
Price (USD)
-
Type:
Adapter for Vial Pusher
Allow the use of 2 mL vials or uncapped 10 mL vials in your Thermo Scientific™ Dionex™ UltiMate™ ACC-3000T. Dionex UltiMate ACC-3000T Vial Adapters add flexibility to your workflow.
Specifications
For Use With (Equipment)Ultimate ACC-3000 T Column Compartment
TypeAdapter for Vial Pusher
Unit SizeEach