TX-400 Rotor Adapters
Thermo Scientific™
TX-400 Rotor Adapters
Use these rotor adapters with round buckets, which enable rotor to accept bottles up to 250 mL and tubes up to 100 mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500370625 mL Universal
7500368076 x 5/7 mL Blood Collection Tube
7500368156 x 10/15 mL Blood Collection Tube
7500368215 mL Conical Tube
7500368350 mL Conical Tube
750037001.5/2.0 mL Conical Microtube
75003788175, 200, 225, 250 mL Bottle
Catalog number 75003706
Price (USD)
-
Capacity:
25 mL Universal
Use Thermo Scientific™ TX-400 Adapters with round buckets in the TX-400 Rotor.
The adapters enable rotor to accept bottles up to 250mL and tubes to 100mL.
  • Maximum adapter capacity: 4 × 250mL (4 × 400mL with 400mL polypropylene bio-bottles)
  • Maximum adapter O.D. × L: 62 × 125mm (80 × 125mm with 400mL bio-bottles)
Specifications
Max. Tube Diameter25 mm
Max. Tube Length110 mm
No. per Pack4
Capacity25 mL Universal
TypeAdapter
Unit SizeEach