TX-750 Swinging Bucket/BIOShield 1000A Adapters
Thermo Scientific™
TX-750 Swinging Bucket/BIOShield 1000A Adapters
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
750037695/7 mL Round-bottom Tube
7500375915 mL Conical Urine Tube
7500376710 mL Blood Collection/15mL DIN Tube
750037685/7 mL Blood Collection Tube
Catalog number 75003769
Price (USD)
-
Capacity:
5/7 mL Round-bottom Tube
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
Specifications
Tube Size5/7 mL
Tube TypeRound Bottom
Capacity5/7 mL Round-bottom Tube
TypeSwinging Bucket Adapter
Unit SizeEach