Manostat™ Norprene™ Pumping Links
Actual product may vary
Thermo Scientific™
Manostat™ Norprene™ Pumping Links
Manostat norprene pumping links are for use with Manostat pumps and dispensers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTubing SizeDescription
7530517517Preston, Spencer and Compulab RD1
7530525525Preston, Spencer and Compulab RD1
7530514514Preston, Spencer and Compulab RD1
7530516516Preston, Spencer and Compulab RD1
7530513513Preston, Spencer and Compulab RD1
7230514514Simon/Vera, Kate and Compulab RD2
7230513513Simon/Vera, Kate and Compulab RD2
7230525525Simon/Vera, Kate and Compulab RD2
7230516516Simon/Vera, Kate and Compulab RD2
7230517517Simon/Vera, Kate and Compulab RD2
Catalog number 75305175
Price (USD)
-
Tubing Size:
17
Description:
Preston, Spencer and Compulab RD1
Thermo Scientific™ Manostat norprene pumping links are for use with Manostat pumps and dispensers.
Specifications
DescriptionPreston, Spencer and Compulab RD1
For Use WithPreston, Spencer and Compulab RD1
Includes5 links/pack
Product LineThermo Scientific
Tubing Size17
Diameter (English) Inner1/4 in.
Diameter (English) Outer0.663 in.
Diameter (Metric) Inner6.4 mm
Diameter (Metric) Outer17 mm
MaterialNorprene
Product TypePumping Link
Unit SizeEach