Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Type | Ammonia Standard Solution, 2 mg/L |
Unit Size | Each |
Type | Ammonia Standard Solution, 5 mg/L |
Unit Size | Each |
Type | Ammonia Standard Solution, 30 mg/L |
Unit Size | Each |
Type | Ammonia Standard Solution, 100 mg/L |
Unit Size | Each |
Type | Ammonia Standard Solution, 500 mg/L |
Unit Size | Each |
Type | 8010cX Annual Maintenance Kit |
Unit Size | Each |
Type | Ammonia Reagent Kit |
Unit Size | Each |
Type | Refrigerator (115 V) for Reagent 2 |
Unit Size | Each |
Type | Refrigerator (220V) for Reagent 2 |
Unit Size | Each |
Current Loops | Two isolated 4-20mA, Max load 900 Ω |
Temperature (Metric) Operating | 5°C to 40°C |
Type | Ammonia Analyzer |
Temperature (English) Operating | 5°C to 40°C |
Length (English) | 26 in. |
Length (Metric) | 660 mm |
Height (English) | 39 in. |
Height (Metric) | 990 mm |
Voltage | 100-240 V |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Type | Price (ARS) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
8010002 | Each | Ammonia Standard Solution, 2 mg/L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010005 | Each | Ammonia Standard Solution, 5 mg/L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010030 | Each | Ammonia Standard Solution, 30 mg/L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010100 | Each | Ammonia Standard Solution, 100 mg/L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010500 | Each | Ammonia Standard Solution, 500 mg/L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010MK | Each | 8010cX Annual Maintenance Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010RE | Each | Ammonia Reagent Kit | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010RFG115 | Each | Refrigerator (115 V) for Reagent 2 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010RFG220 | Each | Refrigerator (220V) for Reagent 2 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
8010CX | Each | Ammonia Analyzer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Orion™8010cX is: