Glass RP-18 W/UV254 TLC Plate

Glass RP-18 W/UV254 TLC Plate

Glass RP-18 W/UV254 TLC Plate
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
8110735 x 20 cm
81107510 x 10 cm
81107420 x 20 cm
Catalog number 811073
Price (USD)
-
Dimensions (L x W):
5 x 20 cm

HPTLC Glass Plates RP-18 W UV254 C18-modified silica layer 5x20cm 0.25mm thick

  • Base material: glass plates with silical gel
  • Mode: Reversed Phase RP-18 W UV254
  • Layer Hardness: Standard, Soft

Adsorbent:

  • Silica Gel with octadecyl (C18, ODS, RP18) modification
  • Phase RP-18 W UV254
  • UV Indicator: Yes
  • Particle Type: Fully porous particles (FPP)
  • Particle Shape: Irregular
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 2-10 μm
  • Binder: Yes

Indicators:

This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).

Binder:

Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Recommended Applications:

Aminophenols, Barbiturates, HPTLC, Nucleobases, Phthalates, Plasticizers, Polycyclic aromatic hydrocarbons (PAH), Preservatives, Reversed phase (RP), Steroids, Tetracyclines, Wettable with water

General TLC Glass Plate Features from Macherey-Nagel:

  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
  • The plate can be easily labelled for clear identification
  • Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
  • Increased separation efficiency due to an optimized particle size distribution
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
Specifications
Dimensions (L x W)5 x 20 cm
Fluorescent IndicatorUV254
For Use With (Application)Aminophenols, Barbiturates, Preservatives, Nucleobases, Polycyclic Aromatic Hydrocarbons, Steroids, Tetracyclines, Plasticizers (Phthalates)
Pore Volume0.75 mL/g
Specific Surface∼500 m2/g
Thickness (Metric)0.25 mm
MaterialGlass RP-18 W
Particle Size2 to 10 μm
Pore Size60 Å
Quantity50 Pk.
TypeTLC Plate
Unit SizeEach