Opti-Link™ Carboxylate-Modified Particles
Thermo Scientific™

Opti-Link™ Carboxylate-Modified Particles

Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)QuantitySurface Acid LoadingConcentration
830009701003900.4 μm100 mLLow Acid10%
730033051002502 μm15 mLHigh Acid10%
730033051003502 μm100 mLHigh Acid10%
730033051004502 μm1000 mLHigh Acid10%
73003420100250
also known as MCG-73003420100250-EACH
3 μm15 mLHigh Acid10%
730034201003503 μm100 mLHigh Acid10%
830005201002900.2 μm15 mLHigh Acid10%
830005201003900.2 μm100 mLHigh Acid10%
830005201004900.2 μm1000 mLHigh Acid10%
830005501002900.2 μm15 mLMedium Acid10%
830005501003900.2 μm100 mLMedium Acid10%
830005501004900.2 μm1000 mLMedium Acid10%
830007201002900.3 μm15 mLHigh Acid10%
830007201003900.3 μm100 mLHigh Acid10%
830007501002900.3 μm15 mLMedium Acid10%
830007501003900.3 μm100 mLMedium Acid10%
830009201002900.4 μm15 mLHigh Acid10%
830009201003900.4 μm100 mLHigh Acid10%
830009701002900.4 μm15 mLLow Acid10%
830009701004900.4 μm1000 mLLow Acid10%
930005701002900.2 μm15 mLLow Acid10%
930005701003900.2 μm100 mLLow Acid10%
930018911002900.85 μm15 mLLow Acid10%
930018911003900.85 μm100 mLLow Acid10%
W004CA0.4 μm15 mLLow Acid4%
W050CA0.5 μm15 mLMedium Acid4%
W050CB0.5 μm100 mLMedium Acid4%
W080CA0.85 μm15 mLMedium Acid4%
W080CB0.85 μm100 mLMedium Acid4%
W090CA0.9 μm15 mLHigh Acid4%
W090CB0.9 μm100 mLHigh Acid4%
W400CA4.0 μm15 mLHigh Acid4%
W400CB4.0 μm100 mLHigh Acid4%
W500CA5.0 μm15 mLHigh Acid4%
W500CB5.0 μm100 mLHigh Acid4%
Catalog number 83000970100390
Price (USD)
-
Diameter (Metric):
0.4 μm
Quantity:
100 mL
Surface Acid Loading:
Low Acid
Concentration:
10%
Control sensitivity, specificity and stability with various acid content available within Thermo Scientific™ Opti-Link™ Carboxylate-Modified Particles. Particles contain carboxylic acid groups for covalent coupling and can be used directly from the bottle in a variety of applications.
  • Parking area combines surface area and acid content to give you the surface acid distribution that is useful for assay optimization
  • Surface area per gram of a particle varies inversely with the particle diameter
  • Acid content refers to the titrated milliequivalents per gram (mEq/g) and is weight-based
  • For most applications, particles can be used directly from the bottle without any pre-cleaning
  • Proprietary anionic surfactant does not interfere with binding of proteins nor cause proteins to desorb from particle surfaces or cause proteins to desorb from particle surfaces
  • Various surface acid concentrations available to help optimize reagent development efforts
  • Particles have been used in and are equally successful in molecular biology and research applications

Parking Area Properties

  • Low Acid Content—60–100 Parking Area—Hydrophobic surface, more colloidally stable than plain polystyrene
  • Medium Acid Content—35–59 Parking Area—Hydrophobic and hydrophilic areas; good colloidal stability and good covalent coupling
  • High Acid Content—10–34 Parking Area—Hydrophilic surface; excellent colloidal stability; excellent covalent coupling

Recommended for:

  • Clinical diagnostics
  • Molecular biology
  • Nephelometric, turbidimetric and slide agglutination assays

Note:

When using Opti-Link particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

Specifications
AdditiveProprietary anionic surfactant
Certifications/ComplianceIncludes Certificate of Analysis
Concentration10%
DescriptionParticles that contain carboxylic acid groups for covalent coupling and that can be used in a variety of applications.
IncludesPackage Insert Sheet containing Certicate of Analysis
Packaging100 mL
Particle Density1.05g/cm³
PreservativePasteurized
Quantity100 mL
Sample Evaluation PacksPackaged in 15mL bottles at 10% solids. The evaluation pack consists of three lots from all available PS or CM particles.
Specialty PackagingOpti-Link Carboxylate-Modified Polystyrene particles are available in different concentrations and package sizes upon request.
Surface Acid LoadingLow Acid
ColorWhite or Colorless
Diameter (Metric)0.4 μm
For Use With (Application)Covalent Coupling
MaterialPolystyrene Sulfate
Product TypeSpecialty Particles
Unit SizeEach
Contents & Storage
Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.