Prohibition Sign - No Entry Authorised Persons Only
Brady Australia

Prohibition Sign - No Entry Authorised Persons Only

Brady Prohibition Signs alert staff of potential hazards in and around the workplace. These highly visible signs are made of self adhesive vinyl so that they can be placed on a variety of surfaces for maximum visualization.
Have Questions?
Catalog NumberMaterial
838563Vinyl
Catalog number 838563
Price (USD)
-
Material:
Vinyl
Brady Prohibition Signs alert staff of potential hazards in and around the workplace. These highly visible signs are made of self adhesive vinyl so that they can be placed on a variety of surfaces for maximum visualization.
  • Alert staff of potential hazards in and around the workplace
  • Black writing on a white sign
  • Made of self adhesive vinyl
  • Dimensions: 250 x 180 mm
Specifications
Displayed TextNo Entry Authorised Persons Only
Height (English)9.8 in.
Height (Metric)250 mm
Size (English)9.8 x 7.1 in.
Size (Metric)250 x 180 mm
Width (English)7.1 in.
Width (Metric)180 mm
ColorWhite
LanguagesEnglish
MaterialVinyl
Mounting TypeSelf-adhesive
ShapeRectangular
Unit SizeEach