IL-27 Human Uncoated ELISA Kit with Plates
Invitrogen™

IL-27 Human Uncoated ELISA Kit with Plates

This Human IL-27 Uncoated ELISA Kit with Plates includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbentRead more
Have Questions?
Catalog NumberQuantity
88-7278-222 x 96 Tests
Catalog number 88-7278-22
Price (USD)
-
Quantity:
2 x 96 Tests
This Human IL-27 Uncoated ELISA Kit with Plates includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays. This kit allows the specific detection of IL-27 with no detection of IL-35 or free p28 or EBI3. This set has been optimized for the accurate and precise measurement of human IL-27 in serum, plasma, and tissue culture supernatant samples. Serum and plasma samples must be diluted 2-fold in prepared assay diluent prior to evaluation in this ELISA.

IL-27 is a member of the IL-12 family, a subgroup of the IL-6 family of cytokines. It is a heterodimer of the subunits EBI3 (Epstein-Barr Virus Induced Gene 3), which is homologous to the p40 subunit shared by IL-12 and IL-23, and p28 (IL-30), which is homologous to p35. IL-27 is produced by activated dendritic cells and macrophages in response to TLR ligands and inflammatory cytokines.

The IL-27 receptor shares one subunit, gp130, with other members of the IL-6 family. The subunit WSX-1 (IL-27Rα, TCCR) is unique to IL-27 and is believed to be the only part of the receptor that interacts with the cytokine. The IL-27R is most abundantly expressed on activated T-cells and NK cells, although expression has also been shown on B-cells and naïve T-cells. IL-27R activation leads to the phosphorylation of Jak/STAT proteins, with STAT1 and STAT3 being critical to the function of IL-27. IL-27 has been shown to have both pro-inflammatory and anti-inflammatory effects. It influences the commitment of CD4+ T-cells toward the Th1 lineage by inducing the expression of the T-bet transcription factor and the upregulation of IL-12Rβ2. Its anti-inflammatory functions include the suppression of Th2 and Th17 proliferation and differentiation. Susceptibility to T-cell mediated autoimmunity has been observed in WSX-1 knockout mice.

Reported Application
ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay Range64-8,000 pg/mL
Assay Sensitivity64 pg/mL
ConjugateBiotin
For Use With (Equipment)Microplate Reader
Former IdentifiersReady-SET-Go!, eBioscience
Geneinterleukin 27
Gene AliasIL-27, IL-27AA, IL27p28, IL30, p28, IL27
Gene ID (Entrez)246778
Gene SymbolIL27
Label or DyeHRP
ProteinInterleukin-27 subunit alpha
Protein AliasesIL-27 subunit alpha, IL-27-A, IL27-A, Interleukin-30, p28
Quantity2 x 96 Tests
Target Kit NamedIL-27
Target Species ValidatedHuman
UniProt IDQ8NEV9
Detection MethodColorimetric
Immunoassay Kit FormatUncoated ELISA Kit with Plates
Target SpeciesHuman
Unit Size2 x 96 tests
Contents & Storage
Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer: 10X PBS Coating Buffer
Assay Diluent: 5X Concentrated
Detection Enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Solution
Certificate of Analysis: Lot-specific instructions for the dilution of antibodies and standards
96-Well Plates: Corning Costar flat-bottom plates (included in catalog numbers ending with -22 and -86)

Frequently asked questions (FAQs)

What are the critical parameters for successful ELISPOT assay?

A: It is critically important to use a high-affinity binding PVDF membrane plate, rather than regular ELISA plate or nitrocellulose membrane plate. Additionally, highest affinity antibodies for capture work best in this assay. Empirical optimization of conditions such as cell density, stimulus/mitogen, and kinetics is necessary.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I recently bought one of your Ready-Set-Go! ELISA Sets, can I use the antibody pairs to run an ELISPOT?

We do not recommend the use of our ELISA kits for the ELISPOT application. We cannot guarantee the successful use of our ELISA kits in an ELISPOT application.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

Do your Ebioscience antibody pairs work in ELISPOT as well?

Most, but not all, of our ELISA antibody pairs work for ELISPOT. We continue to evaluate and optimize our anti-human and anti-mouse cytokine antibody pairs for ELISPOT applications. Please see our protocols page (www.thermofisher.com/us/en/home/references/protocols/cell-and-tissue-analysis/protocols) for a detailed protocol and a reference table with the list of antibody pairs that can be used in ELISPOT.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the components of the Ready-Set-Go! ELISPOT Sets?

Capture Antibody: Pre-titrated, Functional Grade (low endotoxin) purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
10X Sterile Azide-Free Coating Buffer
5X ELISA/ELISPOT Diluent
Detection enzyme: pre-titrated Avidin HRP
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and enzyme

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.