Portavo 902 Cond Basic Conductivity, Resistivity, salinity, TDS Meter
Portavo 902 Cond Basic Conductivity, Resistivity, salinity, TDS Meter
Knick Electronische Messgeraete GmBH
Portavo 902 Cond Basic Conductivity, Resistivity, salinity, TDS Meter
Portavo 902 Cond basic conductivity meter also measures resistivity salinity and TDS with M8 socket for Memosens sensors and analog DIN socket plus
Have Questions?
Catalog NumberIncludes
902CONDM8 socket
Catalog number 902COND
Price (USD)
-
Includes:
M8 socket
Portavo 902 Cond basic conductivity meter also measures resistivity salinity and TDS with M8 socket for Memosens sensors and analog DIN socket plus
  • Portavo 902 Cond basic conductivity meter
  • Measures resistivity salinity and TDS
  • With M8 socket
  • For Memosens sensors and analog DIN socket plus 2 x 4 mm for temp
Specifications
Accuracy (Conductivity)<0.5 % meas.val. + 0.4 μS
For Use With (Application)Memosens Sensors and Analog DIN Socket
IncludesM8 socket
Display Type7-segment LCD STN with 3 lines and Icons
IP RatingIP 66/67 with pressure compensation
Measurement ModesConductivity, Resistivity, Salinity, TDS
Temperature Range (Metric)-10°C to +55°C
Unit SizeEach